Learn More
MilliporeSigma™ Calbiochem™ BAPTA/AM
Can be loaded into wide variety of cells, where it is hydrolyzed by cytosolic esterases and is trapped intracellularly as the active chelator BAPTA
Supplier:  MilliporeSigma™ 196419100MG
 View more versions of this product
Catalog No. 196419100MG
Only null left
Explore available promotions
Description
Description
  • Membrane-permeable form of BAPTA
  • Prevents cocaine-induced ventricular fibrillations
  • Abolishes vitamin D3-induced increase in intracellular Ca2+
  • Induces inactivation of protein kinase C
  • Also inhibits thapsigargin-induced apoptosis in rat thymocytes
Specifications
Specifications
Y
126150-97-8
Solid
1,2-bis(o-Aminophenoxy)ethane-N,N,Nʹ,Nʹ-tetraacetic Acid Tetra(acetoxymethyl) Ester
Calbiochem BAPTA/AM
≥90% by TLC
-20°C
100 mg
DMSO
Product Suggestions
Product Suggestions