missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ BAPTA/AM

Catalog No. p-6814337
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19641925MG 25 mg
196419100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19641925MG Supplier MilliporeSigma™ Supplier No. 19641925MG
Only null left

Can be loaded into wide variety of cells, where it is hydrolyzed by cytosolic esterases and is trapped intracellularly as the active chelator BAPTA

  • Membrane-permeable form of BAPTA
  • Prevents cocaine-induced ventricular fibrillations
  • Abolishes vitamin D3-induced increase in intracellular Ca2+
  • Induces inactivation of protein kinase C
  • Also inhibits thapsigargin-induced apoptosis in rat thymocytes

Specifications

Membrane Permeable Y
Purity ≥90% by TLC
CAS 126150-97-8
Content And Storage -20°C
Form Solid
Quantity 25 mg
Synonym 1,2-bis(o-Aminophenoxy)ethane-N,N,Nʹ,Nʹ-tetraacetic Acid Tetra(acetoxymethyl) Ester
Solubility DMSO
Product Type Calbiochem BAPTA/AM