missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Aprotinin, Bovine, Recombinant, Nicotiana sp., Animal-Free

Aprotinin, Bovine, Recombinant, Nicotiana sp, Animal-Free - CAS 9087-70-1, is a heat and acid-stable competitive, reversible inhibitor of serine proteases.

$122.38 - $386.61

Specifications

Color Clear Colorless to Amber
Formulation In 0.9% NaCl. Contains no animal-derived components.
Target Proteins chymotrypsin
CAS 9087-70-1
Molecular Formula C284H432N84O79S7
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6163711MG
View Documents
MilliporeSigma™
6163711MG
1 mg
Each for $122.38
Only null left
 
61-637-15MG
View Documents
MilliporeSigma™
6163715MG
5 mg
Each for $213.62
Only null left
 
61-637-125MG
View Documents
MilliporeSigma™
61637125MG
25 mg
Each for $386.61
Only null left
 
Description

Description

Recombinant, bovine aprotinin supplied without animal-derived components. Aprotinin is a competitive, reversible inhibitor of proteolytic and esterolytic activity. A relatively heat- and acid-stable serine protease inhibitor. Forms a tight complex, blocking the active site of the enzyme. Effective at concentrations equimolar with protease. Inhibits several proteases, including coagulation factors in the prephase of blood clotting, tissue and leukocytic proteinases, chymotrypsin, trypsin (Kd = 5 x 10-14 M), plasmin (Kd = 2.3 x 10-10 M), and kallikrein (Kd = 1 x 10-7 M). Proteases not inhibited by aprotinin include Factor Xa, thrombin, pepsin, papain, and carboxypeptidases A and B. Useful for protein purification and for extending the life of cells in culture by preventing proteolytic damage.
Specifications

Specifications

Clear Colorless to Amber
chymotrypsin
C284H432N84O79S7
Liquid
In 0.9% NaCl. Contains no animal-derived components.
9087-70-1
6512g/mol
95% by SDS-PAGE
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'