Learn More
MilliporeSigma™ Calbiochem™ Aprotinin, Bovine, Recombinant, Nicotiana sp., Animal-Free
Aprotinin, Bovine, Recombinant, Nicotiana sp, Animal-Free - CAS 9087-70-1, is a heat and acid-stable competitive, reversible inhibitor of serine proteases.
Supplier:  MilliporeSigma™ 61637125MG
 View more versions of this product
Catalog No. 61-637-125MG
Only null left
Explore available promotions
Description
Description
Recombinant, bovine aprotinin supplied without animal-derived components. Aprotinin is a competitive, reversible inhibitor of proteolytic and esterolytic activity. A relatively heat- and acid-stable serine protease inhibitor. Forms a tight complex, blocking the active site of the enzyme. Effective at concentrations equimolar with protease. Inhibits several proteases, including coagulation factors in the prephase of blood clotting, tissue and leukocytic proteinases, chymotrypsin, trypsin (Kd = 5 x 10-14 M), plasmin (Kd = 2.3 x 10-10 M), and kallikrein (Kd = 1 x 10-7 M). Proteases not inhibited by aprotinin include Factor Xa, thrombin, pepsin, papain, and carboxypeptidases A and B. Useful for protein purification and for extending the life of cells in culture by preventing proteolytic damage.
Specifications
Specifications
Clear Colorless to Amber
chymotrypsin
C284H432N84O79S7
Liquid
95% by SDS-PAGE
In 0.9% NaCl. Contains no animal-derived components.
9087-70-1
6512g/mol
25 mg
Product Suggestions
Product Suggestions