Learn More
MilliporeSigma™ Calbiochem™ Phorbol-12-myristate-13-acetate
Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.
Supplier: MilliporeSigma™ 52440025MG