missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Phorbol-12-myristate-13-acetate

Catalog No. p-7060122
Change view
Click to view available options
Quantity:
25 mg
5 mg
Catalog No. Quantity
5244005MG 5 mg
52440025MG 25 mg
2 options

Catalog No. 5244005MG

Supplier: MilliporeSigma™ 5244005MG

Only null left

Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.

Packaged Under Inert Gas

Specifications

Color White
Target Proteins Protein Kinase C
CAS 16561-29-8
Molecular Formula C36H56O8
Formula Weight 616.8g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Purity 98% by HPLC