Learn More
MilliporeSigma™ Calbiochem™ Pepstatin A, Synthetic
Pepstatin A, Synthetic, CAS 26305-03-3, is a reversible inhibitor of aspartic proteases. Inhibits cathepsin D, pepsin, and renin.
Supplier:  MilliporeSigma™ 51648125MG
 View more versions of this product
Catalog No. 51-648-125MG
Only null left
Explore available promotions
Description
Description
A reversible inhibitor of aspartic proteases. Inhibits cathepsin D, pepsin, and renin. Reported to block apoptosis induced by CA-074 (Cat. No. 205530) in PC12 cell lines. Also available as a 25mm solution in DMSO (Cat. No. 508437).
Specifications
Specifications
White
26305-03-3
685.9g/mol
Solid
98% by HPLC
cathepsin D
C34H63N5O9
DMSO or MeOH
25 mg
Product Suggestions
Product Suggestions