missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Pepstatin A, Synthetic

Catalog No. p-7060118
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5164815MG 5 mg
51648125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5164815MG Supplier MilliporeSigma™ Supplier No. 5164815MG
Only null left

Pepstatin A, Synthetic, CAS 26305-03-3, is a reversible inhibitor of aspartic proteases. Inhibits cathepsin D, pepsin, and renin.

A reversible inhibitor of aspartic proteases. Inhibits cathepsin D, pepsin, and renin. Reported to block apoptosis induced by CA-074 (Cat. No. 205530) in PC12 cell lines. Also available as a 25mm solution in DMSO (Cat. No. 508437).

Specifications

Color White
Target Proteins cathepsin D
CAS 26305-03-3
Molecular Formula C34H63N5O9
Formula Weight 685.9g/mol
Solubility DMSO or MeOH
Form Solid
Quantity 5 mg
Purity 98% by HPLC