missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ LY 294002
LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 μM). Acts on the ATP-binding site.
$299.26 - $299.26
Specifications
| Color | Off-white |
|---|---|
| Target Proteins | Phosphatidylinositol 3-kinase |
| CAS | 154447-36-6 |
| Molecular Formula | C19H17NO3 |
| Formula Weight | 307.4g/mol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
44-020-25MG
|
MilliporeSigma™
4402025MG |
5 mg |
Each for $299.26
|
|
|||||
Description
Cell PermeableSpecifications
| Off-white | |
| 154447-36-6 | |
| 307.4g/mol | |
| Protect from light | |
| 98% by HPLC |
| Phosphatidylinositol 3-kinase | |
| C19H17NO3 | |
| DMSO or EtOH | |
| Solid |