missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Autophagy Inhibitor, 3-MA
Autophagy Inhibitor, 3-MA, CAS 5142-23-4, is a cell-permeable autophagic sequestration blocker. Acts as an inhibitor of III PI3-Kinase.
$216.98 - $216.98
Specifications
| Color | White to Faint Yellow |
|---|---|
| CAS | 5142-23-4 |
| Molecular Formula | C6H7N5 |
| Formula Weight | 149.2g/mol |
| Solubility | DMSO or H2O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-949-0100MG
|
MilliporeSigma™
189490100MG |
100 mg |
Each for $216.98
|
|
|||||
Description
Packaged Under Inert GasSpecifications
| White to Faint Yellow | |
| C6H7N5 | |
| DMSO or H2O | |
| Powder |
| 5142-23-4 | |
| 149.2g/mol | |
| Protect from light | |
| 95% by HPLC |