missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, N6,O2'-Dibutyryl-, Sodium Salt

This Dibutyryl-cAMP, CAS 16980-89-5, is a cell-permeable cAMP analog that preferentially activates PKA.

$614.77 - $614.77

Specifications

Color White
Target Proteins PKA
CAS 16980-89-5
Molecular Formula C18H23N5O8P·Na
Formula Weight 491.4g/mol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
28-745-100MG
View Documents
MilliporeSigma™
28745100MG
100 mg
Each for $614.77
Only null left
 
Description

Description

Cell Permeable
Specifications

Specifications

White
16980-89-5
491.4g/mol
Hygroscopic
95% by HPLC
PKA
C18H23N5O8P·Na
H2O
Lyophilized
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'