CAS RN 16980-89-5
IUPAC Name:
sodium 6-(6-butanamido-9H-purin-9-yl)-7-(butanoyloxy)-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonyms:
sodium 6-(6-butanamidopurin-9-yl)-7-(butanoyloxy)-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Weight (g/mol):
491.37
Molecular Formula:
C18H23N5NaO8P
InChi Key:
KRBZRVBLIUDQNG-UHFFFAOYNA-M
SMILES:
[Na+].CCCC(=O)NC1=C2N=CN(C3OC4COP([O-])(=O)OC4C3OC(=O)CCC)C2=NC=N1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, N6,O2'-Dibutyryl-, Sodium Salt
This Dibutyryl-cAMP, CAS 16980-89-5, is a cell-permeable cAMP analog that preferentially activates PKA.
2
N6,2'-O-Dibutyryladenosine 3',5'-Cyclic Monophosphate Sodium Salt 97.0+%, TCI America™
CAS: 16980-89-5 Molecular Formula: C18H26N5NaO8P Molecular Weight (g/mol): 494.397 MDL Number: MFCD00005843 InChI Key: KSSDVZRQZQECKZ-VMZSDVDQSA-N Synonym: bucladesine PubChem CID: 131673831 IUPAC Name: [(4aR,6R,7R,7aR)-6-[6-(butanoylamino)purin-9-yl]-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-yl] butanoate;molecular hydrogen;sodium SMILES: [HH].CCCC(=O)NC1=NC=NC2=C1N=CN2C3C(C4C(O3)COP(=O)(O4)O)OC(=O)CCC.[Na]