missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ 5-Bromo-2'-deoxyuridine

Can be used both in vitro and in vivo to label DNA during DNA synthesis where it is incorporated into DNA in place of thymidine

Specifications

CAS 59-14-3
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 307.1g/mol
Purity ≥98% by HPLC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
2038065GM
View Documents Promotion Details
MilliporeSigma™
2038065GM
5 g N/A
Only null left
 
2038061GM
View Documents Promotion Details
MilliporeSigma™
2038061GM
1 g N/A
Only null left
 
Description

Description

  • Thymidine analog
  • Study of DNA synthesis where it is incorporated into DNA in place of thymidine
  • Can be used both in vitro and in vivo to label DNA
  • The resulting DNA can be used in conjunction with BrdU (Ab-2) Monoclonal Antibody for immunohistochemical analysis of cell proliferation
Specifications

Specifications

59-14-3
Store at +15°C to +30°C
≥98% by HPLC
Solid
White
307.1g/mol
5-BrdU
Videos
SDS
Documents

Documents