missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ 5-Bromo-2'-deoxyuridine

Catalog No. 2038065GM
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2038065GM 5 g
2038061GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2038065GM Supplier MilliporeSigma™ Supplier No. 2038065GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Can be used both in vitro and in vivo to label DNA during DNA synthesis where it is incorporated into DNA in place of thymidine

  • Thymidine analog
  • Study of DNA synthesis where it is incorporated into DNA in place of thymidine
  • Can be used both in vitro and in vivo to label DNA
  • The resulting DNA can be used in conjunction with BrdU (Ab-2) Monoclonal Antibody for immunohistochemical analysis of cell proliferation

Specifications

CAS 59-14-3
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 307.1g/mol
Purity ≥98% by HPLC
Quantity 5 g
Synonym 5-BrdU
Form Solid