missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ 5-Bromo-2'-deoxyuridine

Can be used both in vitro and in vivo to label DNA during DNA synthesis where it is incorporated into DNA in place of thymidine

Supplier:  MilliporeSigma™ 2038061GM

 View more versions of this product

Catalog No. 2038061GM


Only null left

Description

Description

  • Thymidine analog
  • Study of DNA synthesis where it is incorporated into DNA in place of thymidine
  • Can be used both in vitro and in vivo to label DNA
  • The resulting DNA can be used in conjunction with BrdU (Ab-2) Monoclonal Antibody for immunohistochemical analysis of cell proliferation
Specifications

Chemical Identifiers

59-14-3
Store at +15°C to +30°C
≥98% by HPLC
Solid
White
307.1g/mol
5-BrdU

Specifications

59-14-3
Store at +15°C to +30°C
≥98% by HPLC
5-BrdU
White
307.1g/mol
1 g
Solid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions