missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CA19-9 ELISA Kit

ELISA

$900.00 - $4,420.00

Spécifications

Assay Range 0.274 to 200 U/mL
Assay Sensitivity 0.3 U/mL
Calibration Range 0.3 U/mL to 200 U/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
EHCA199
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
EHCA199
96 Tests
paquet for $900.00
Il en reste null
 
EHCA199X5
Afficher les documents Détails de la promotion
Invitrogen™
EHCA199X5
5 x 96 Tests
paquet de 5 chaque for $4,420.00
Il en reste null
 
Description

Description

The Human CA19-9 ELISA quantitates Hu CA19-9 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CA19-9. Principle of the method The Human CA19-9 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Mucin glycoprotein is a Sialyl Lewis A structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewis A and/or Lewis B blood group antigens. In normal tissues, sialyl Lewis A antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas, including adenocarcinomas of the stomach, intestine, and pancreas. Preoperative elevated CA19-9 levels in patients with stage I pancreatic carcinoma decrease to normal values following surgery. When used serially, CA19-9 can predict recurrence of disease prior to radiographic or clinical findings.
Spécifications

Spécifications

0.274 to 200 U/mL
0.3 U/mL to 200 U/mL
HRP
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF-7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP- 2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
CA19-9
4 hrs 45 mins
0.3 U/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
<10
HRP
Other Research
2°C to 8°C
1 hrs 20 mins
Videos
FDS
Documents

Documents