missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ RT Series Supplied-Air Respirator Breathing Tubes

Numéro de catalogue. 361003802
Change view
Click to view available options
Description:
Lightweight, disposable breathing tube
Wire-reinforced, heavy-duty breathing tube
Color:
Gray
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Description Color
361003802 Wire-reinforced, heavy-duty breathing tube White
361003803 Lightweight, disposable breathing tube Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 361003802 Fournisseur Bullard™ Code fournisseur 20BT
Il en reste null

Breathing tube component for use with RT Series respirators.

  • Component for use with RT series respirators
  • Choose between heavy-duty or lightweight options
  • Breathing tubes must be used in conjunction with hood, flow control device, and air hose

Spécifications

Description Wire-reinforced, heavy-duty breathing tube
Color White
Product Type Breathing Tube
For Use With (Equipment) RT Series supplied-air respirators
Includes S18051 hose clamp
Model 20BT