missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Model 88VX Respirator

Catalog No. 19088314A
Change view
Click to view available options
Certifications/Compliance:
NIOSH C or CE, OSHA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
19088314A NIOSH C or CE, OSHA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19088314A Supplier Bullard™ Supplier No. 88VX3230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Helmet and 28 in. long (71cm) medium-weight cape to protect wearer's entire head, face, and upper body from abrasive rebound, airborne contaminants, and messy overspray
  • Also comes with breathing tube assembly and flow control valve, 1/4 in. Industrial Interchange fitting and belt, and inner and outer lenses; order breathing air supply hoses separately
  • Durable breathing tube for compressed breathing air includes a coil-spring reinforced breathing tube, constant or adjustable airflow control, and belt

Provides comfortable, reliable respiratory protection for abrasive blasters and painters.

  • Lightweight inner shell with adjustable headband holds respirator firmly and comfortably on wearer's head
  • Wide-angle window contains both an inner and an outer lens positioned for optimal visibility
  • Outer lens protects main inner lens from abrasion

Certifications

NIOSH approved as Type C or CE Continuous Flow Class Respirator

Compliance

  • Helmet meets ANSI Z89.1-1969 Class A (except weight) requirements for top impact and penetration
  • Inner lens meets ANSI Z87.1-1989 for impact and penetration protection

Specifications

Certifications/Compliance NIOSH C or CE, OSHA
Color Yellow
Material Polyethylene
Product Type Airline Respirator
Size Standard
For Use With (Equipment) Air respiratory system
Includes Helmet, 28 in. long (71cm) medium-weight cape, breathing tube assembly, flow control valve, 1/4 in. Industrial Interchange fitting and belt, and inner and outer lenses
Model 88VX™
Length (English) 28 in.
Length (Metric) 71.12 cm