missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Bulk Pack of Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
This bulk pack of narrow-mouth translucent HDPE reagent bottles with polypropylene caps is leak-tested, chemical-resistant, and ideal for high-volume lab or industrial use where reliable liquid handling and visibility matter.
$896.96 - $1,973.36
Specifications
| Color | Translucent |
|---|---|
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000104
|
Eisco Labs
CH5412PK2000 |
8 mL |
Pack of 2000 Each for $1,973.36
|
|
|||||
|
S000105
|
Eisco Labs
CH5414PK1000 |
30 mL |
Pack of 1000 Each for $1,614.57
|
|
|||||
|
S000106
|
Eisco Labs
CH5416PK500 |
125 mL |
Pack of 500 Each for $1,614.57
|
|
|||||
|
S000107
|
Eisco Labs
CH5417PK250 |
250 mL |
Pack of 250 Each for $1,291.64
|
|
|||||
|
S000108
|
Eisco Labs
CH5418PK125 |
500 mL |
Pack of 125 Each for $896.96
|
|
|||||
Description
- Bulk pack of reagent bottles - Includes narrow mouth translucent HDPE bottles with polypropylene caps. Ideal for large-scale lab or industrial applications.
- Chemical resistant bottles - Constructed from HDPE for excellent resistance to a variety of chemicals, making these bottles versatile for storing reagents, solutions, and samples.
- Leak-proof tested for reliability - Each bottle is rigorously tested to ensure no leaks under pressure. Perfect for safe storage and transport of liquids in laboratory and industrial settings.
- Lab-friendly features - Narrow mouth for controlled pouring. Translucent design allows easy monitoring of liquid levels during use.
- Suited for extreme conditions - Durable and reliable with a temperature range of -100°C to +120°C. Perfect for diverse applications in laboratories, research facilities, and industrial environments.
Specifications
| Translucent | |
| Non-sterile | |
| No | |
| Reagent Bottle |
| HDPE | |
| Yes | |
| Narrow | |
| Not Autoclavable |