missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Bulk Pack of Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
Description
- Bulk pack of reagent bottles - Includes narrow mouth translucent HDPE bottles with polypropylene caps. Ideal for large-scale lab or industrial applications.
- Chemical resistant bottles - Constructed from HDPE for excellent resistance to a variety of chemicals, making these bottles versatile for storing reagents, solutions, and samples.
- Leak-proof tested for reliability - Each bottle is rigorously tested to ensure no leaks under pressure. Perfect for safe storage and transport of liquids in laboratory and industrial settings.
- Lab-friendly features - Narrow mouth for controlled pouring. Translucent design allows easy monitoring of liquid levels during use.
- Suited for extreme conditions - Durable and reliable with a temperature range of -100°C to +120°C. Perfect for diverse applications in laboratories, research facilities, and industrial environments.
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Capacity (English) | 1 oz. |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 14 mm |
| Material | HDPE |
| No. per Pack | 1000 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Narrow |
| Show More |