Learn More
Eisco Labs Bulk Pack of Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
This bulk pack of narrow-mouth translucent HDPE reagent bottles with polypropylene caps is leak-tested, chemical-resistant, and ideal for high-volume lab or industrial use where reliable liquid handling and visibility matter.
Supplier:  Eisco Labs CH5417PK250
 View more versions of this product
Catalog No. S000107
Only null left
Description
Description
  • Bulk pack of reagent bottles - Includes narrow mouth translucent HDPE bottles with polypropylene caps. Ideal for large-scale lab or industrial applications.
  • Chemical resistant bottles - Constructed from HDPE for excellent resistance to a variety of chemicals, making these bottles versatile for storing reagents, solutions, and samples.
  • Leak-proof tested for reliability - Each bottle is rigorously tested to ensure no leaks under pressure. Perfect for safe storage and transport of liquids in laboratory and industrial settings.
  • Lab-friendly features - Narrow mouth for controlled pouring. Translucent design allows easy monitoring of liquid levels during use.
  • Suited for extreme conditions - Durable and reliable with a temperature range of -100°C to +120°C. Perfect for diverse applications in laboratories, research facilities, and industrial environments.
Specifications
Chemical Identifiers
Translucent
Non-sterile
No
Reagent Bottle
HDPE
Yes
Narrow
Not Autoclavable
Specifications
250 mL
Translucent
HDPE
Non-sterile
No
133 mm
61.7 mm
8.45 oz.
17.6 mm
250 Pk.
Yes
Narrow
Reagent Bottle
Not Autoclavable
Product Suggestions
Product Suggestions