missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Bulk Pack of Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps

Catalog No. S000104
Change view
Click to view available options
Capacity (Metric):
8 mL
30 mL
125 mL
250 mL
500 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000104 8 mL
S000105 30 mL
S000106 125 mL
S000107 250 mL
S000108 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S000104 Supplier Eisco Labs Supplier No. CH5412PK2000
Only null left

This bulk pack of narrow-mouth translucent HDPE reagent bottles with polypropylene caps is leak-tested, chemical-resistant, and ideal for high-volume lab or industrial use where reliable liquid handling and visibility matter.

  • Bulk pack of reagent bottles - Includes narrow mouth translucent HDPE bottles with polypropylene caps. Ideal for large-scale lab or industrial applications.
  • Chemical resistant bottles - Constructed from HDPE for excellent resistance to a variety of chemicals, making these bottles versatile for storing reagents, solutions, and samples.
  • Leak-proof tested for reliability - Each bottle is rigorously tested to ensure no leaks under pressure. Perfect for safe storage and transport of liquids in laboratory and industrial settings.
  • Lab-friendly features - Narrow mouth for controlled pouring. Translucent design allows easy monitoring of liquid levels during use.
  • Suited for extreme conditions - Durable and reliable with a temperature range of -100°C to +120°C. Perfect for diverse applications in laboratories, research facilities, and industrial environments.

Specifications

Capacity (Metric) 8 mL
Capacity (English) 0.27 oz.
Color Translucent
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Pack 2000 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 45 mm
Product Type Reagent Bottle
Diameter (Metric) 25 mm
Autoclavable Not Autoclavable
Show More Show Less