Learn More
Cytiva Buffer Valve
Description
For use with the AKTA start automated protein purification system. The buffer valve is used as a switching valve for gradient formation. It enables the use of two buffers, A and B, which are required for forming gradient during runs.
Spécifications
Spécifications
| For Use With (Application) | Used as a switching valve for gradient formation |
| For Use With (Equipment) | AKTA start Automated Protein Purification System |