missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 6.00 (Certified), Fisher Chemical™

Numéro de catalogue. p-213039
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
SB104500 500 mL Poly Bottle
SB1041 1 L Poly Bottle
SB10420 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. SB104500 Fournisseur Thermo Fisher Scientific Code fournisseur FLSB104500
Il en reste null

Spécifications

Chemical Name or Material Buffer Solution, pH 6.00
Melting Point 0°C
CAS 7778-77-0, 7732-18-5, 1310-73-2
Boiling Point 100°C
Color Undesignated
pH 6.00
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Afficher plus de résultats Afficher moins de résultats