missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 3.5 R, Ricca Chemical

Numéro de catalogue. 14748
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
14748 250 mL
147416 500 mL
147432 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 14748 Fournisseur Ricca Chemical Company Code fournisseur 14748
Il en reste null

Spécifications

Chemical Name or Material Buffer Solution
Name Note pH 3.5 Reagent
CAS 7647-01-0, 631-61-8, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 3.5
Physical Form Liquid
Quantity 250 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Afficher plus de résultats Afficher moins de résultats