missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 1.0 at 25°C, Certified, LabChem™

Supplier:  LabChem LC121802

 View more versions of this product

Catalog No. LC121802


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Buffer Solution pH 1.0 at 25°C
Colorless
Chlorine; Thermal decomposition generates: Corrosive vapors
1.00 ±0.02
1 L
KCl
UN1789
Traceable to NIST
74.55
99.58,0.37,0.05
7447-40-7 , 7647-01-0
1g/mL
Liquid
Passes Test
Poly Bottle
1g/mL
Soluble in water
Certified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions