missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 1.0 at 25°C, Certified, LabChem™

Catalog No. p-5379468
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC121801 500 mL
LC121802 1 L
LC121804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC121801 Supplier LabChem Supplier No. LC121801
Only null left

Specifications

Chemical Name or Material Buffer Solution pH 1.0 at 25°C
Color Colorless
CAS Max % 99.58, 0.37, 0.05
CAS 7447-40-7, 7647-01-0
Decomposition Information Chlorine; Thermal decomposition generates: Corrosive vapors
Density 1g/mL
pH 1.00 ±0.02
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula KCl
Packaging Poly Bottle
UN Number UN1789
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 74.55
Grade Certified
Show More Show Less