missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 7.40 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 156532

 View more versions of this product

Catalog No. 36-101-8155


Only null left

Description

Description

Specifications

Specifications

Buffer, Reference Standard
7558-79-4 , 7778-77-0 , 7732-18-5
Colorless
1X
pH Meter and Electrode
Liquid
pH Buffer
pH 7.40± 0.01 at 25°C
97.0,0.86,0.28
100.0,0.9,0.29
Calibration
7.4
1 L
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions