missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 15501CT

 View more versions of this product

Catalog No. 15501CT


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Buffer, Reference Standard
7558-79-4 , 7778-77-0 , 7732-18-5
100.0,0.91,0.5
Calibration
7
4 L
Cubitainer
pH 7.00 ± 0.01 at 25 deg C
97.0,0.87,0.48
1X
pH Meter and Electrode
Liquid
pH Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions