missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 3.56 ± 0.01 at 25°C, Ricca Chemical
$76.68 - $103.57
Specifications
| Chemical Name or Material | Buffer, Reference Standard |
|---|---|
| Name Note | pH 3.56± 0.01 at 25°C |
| CAS | 877-24-7 , 7647-01-0 , 7732-18-5 |
| CAS Min % | 97.0,1.0,0.06 |
| CAS Max % | 100.0,1.04,0.06 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
36-101-8130
|
Ricca Chemical Company
149816 |
500 mL | Natural Poly Bottle |
Each for $76.68
|
|
||||
|
36-101-8131
|
Ricca Chemical Company
149832 |
1 L | Natural Poly Bottle |
Each for $103.57
|
|
||||