missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 3.56 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 149832

 View more versions of this product

Catalog No. 36-101-8131


Only null left

Description

Description

Specifications

Specifications

Buffer, Reference Standard
877-24-7 , 7647-01-0 , 7732-18-5
Colorless
1X
pH Meter and Electrode
Liquid
pH Buffer
Non-sterile
pH 3.56± 0.01 at 25°C
97.0,1.0,0.06
100.0,1.04,0.06
Calibration
3.56
1 L
Natural Poly Bottle
Reference Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions