missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 13.00 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 16255

 View more versions of this product

Catalog No. 16255


Only null left

Description

Description

Specifications

Specifications

Buffer, Reference Standard
7447-40-7 , 7732-18-5 , 7447-40-7
Colorless
1X
pH Meter and Electrode
Liquid
pH Buffer
pH 13.00± 0.01 at 25°C
97.0,0.91,0.36
100.0,0.95,0.37
Calibration
13
20 L
Cubitainer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions