missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 13.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. p-3251424
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Clear PET-G Bottle
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
162516 500 mL Clear PET-G Bottle
162532 1 L Natural Poly Bottle
16251 4 L Natural Poly Bottle
162525 10 L Cubitainer
16255 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 162516 Supplier Ricca Chemical Company Supplier No. 162516
Only null left

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 13.00± 0.01 at 25°C
CAS 7447-40-7, 7732-18-5, 7447-40-7
CAS Min % 97.0, 0.91, 0.36
Color Colorless
CAS Max % 100.0, 0.95, 0.37
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 13
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Clear PET-G Bottle
Show More Show Less