missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 160025

 View more versions of this product

Catalog No. 1600-25


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Buffer, Reference Standard
497-19-8 , 144-55-8 , 1310-73-2 , 7732-18-5
100.0,0.53,0.41,0.03
Calibration
10
10 L
Cubitainer
pH 10.00 ± 0.01 at 25 deg C
97.0,0.51,0.39,0.03
1X
pH Meter and Electrode
Liquid
pH Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions