missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 1.07 ± 0.01 at 25°C, Ricca Chemical

$72.14 - $313.87

Specifications

CAS 7647-01-0 , 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
36-101-8122
View Documents
Ricca Chemical Company
149016
500 mL Natural Poly Bottle
Each for $72.14
Only null left
 
36-101-8123
View Documents
Ricca Chemical Company
149032
1 L Natural Poly Bottle
Each for $114.89
Only null left
 
1490-1
View Documents
Ricca Chemical Company
14901
4 L Natural Poly Bottle
Each for $244.07
Only null left
 
1490-25
View Documents
Ricca Chemical Company
149025
10 L Cubitainer
Each for $313.87
Only null left
 
Description

Description

Specifications

Specifications

7647-01-0 , 7732-18-5
1X
pH Meter and Electrode
Liquid
Colorless
Calibration
1.07
pH Buffer
Videos
SDS
Documents

Documents