missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 1.07 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 149016

 View more versions of this product

Catalog No. 36-101-8122


Only null left

Description

Description

Specifications

Specifications

Buffer, Reference Standard
7647-01-0 , 7732-18-5
1X
pH Meter and Electrode
Liquid
pH Buffer
pH 1.07± 0.01 at 25°C
Colorless
Calibration
1.07
500 mL
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions