missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genetrodes™ Electrodes
Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.
Supplier: BTX™ 45 0115
Description
- Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Five models available
- Models 508 and 510 are straight with gold tips designed for general in vivo applications
- Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation
Order Info
Genetrode holder with shaft available separately (Cat. No. BTX515).
Identifiants chimiques
| In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery | |
| 200 V | |
| ∼1500+ Pulses | |
| 10 μsec. to 99 msec. in PBS | |
| 0.03 to 0.39 in. | |
| Gold Tip | |
| Banana Plug to Micrograbber |
| Gold | |
| Genetrode only | |
| 1 sec. to 99 msec. | |
| 1 Ea. | |
| 1 to 10 mm | |
| Non-sterile | |
| Square Wave |
Spécifications
| In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery | |
| Gold | |
| Electroporation accessory | |
| Genetrodes, 5 mm L-Shape (GOLD TIP) | |
| Genetrode only | |
| 5 mm | |
| 1 sec. to 99 msec. | |
| 1 Ea. | |
| 1 to 10 mm | |
| Non-sterile | |
| Square Wave |
| ECM 830, ECM 2001+, ECM 2001, and Gemini X2 | |
| BTX Electroporators | |
| 200 V | |
| Bent L-shaped | |
| 0.19 in. | |
| ∼1500+ Pulses | |
| 10 μsec. to 99 msec. in PBS | |
| 0.03 to 0.39 in. | |
| Gold Tip | |
| Banana Plug to Micrograbber |
Intended for Research use only. Not for use in diagnostic or clinical procedures.