missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genetrodes™ Electrodes
Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.
$1,353.04
Specifications
| For Use With (Application) | In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery |
|---|---|
| Material | Gold |
| Voltage | 200 V |
| Includes | Genetrode only |
| Life Span | ∼1500+ Pulses |
| Catalog Number | Mfr. No. | Electrode Shape | Length (Metric) Electrode | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Electrode Shape | Length (Metric) Electrode | Price | Quantity & Availability | ||||
|
BTX514
|
BTX™
45 0116 |
Bent L-shaped | 3 mm |
Each for $1,353.04
|
|
||||
|
BTX512
|
BTX™
45 0115 |
Bent L-shaped | 5 mm |
Each for $1,353.04
|
|
||||
|
BTX516
|
BTX™
45 0117 |
Bent L-shaped | 1 mm |
Each for $1,353.04
|
|
||||
|
BTX510
|
BTX™
45 0114 |
Straight | 10 mm |
Each for $1,353.04
|
|
||||
Description
- Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Five models available
- Models 508 and 510 are straight with gold tips designed for general in vivo applications
- Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation
Order Info
Genetrode holder with shaft available separately (Cat. No. BTX515).
Specifications
| In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery | |
| 200 V | |
| ∼1500+ Pulses | |
| 10 μsec. to 99 msec. in PBS | |
| 0.03 to 0.39 in. | |
| Gold Tip | |
| Banana Plug to Micrograbber |
| Gold | |
| Genetrode only | |
| 1 sec. to 99 msec. | |
| 1 Ea. | |
| 1 to 10 mm | |
| Non-sterile | |
| Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.