missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTEX Solution, MilliporeSigma™ Supelco™
Description
Environmental
Spécifications
Spécifications
| CAS | 108-88-3 |
| Density | 1.099 g/mL (at 20°C) |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Grade | Analytical Standard |