missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent
Description
Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.
Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.
- Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
- Catalyzed formulation is stronger than BSTFA alone
Specifications
Specifications
| For Use With | GC |
| Standard Concentration | 98 to 100, 0.1 to 1% |
| Storage | 2°C to 8°C |
| Quantity | 100 g |
| Standard Components | BSTFA +1% TMCS |
For Research Use Only. Not for use in diagnostic procedures.