missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. p-7157070
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601097 100 mL Poly Bottle
18601098 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601097 Supplier Spectrum Chemical Mfg Cor Supplier No. B303100ML
Only null left

B-303, 115-39-9

Spectrum™ Chemical Bromophenol Blue TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 62625-28-9 , 7732-18-5 , 64-17-5
Molecular Formula C19H9Br4NaO5S
Molecular Weight (g/mol) 691.94
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
IUPAC Name sodium 2-[(3,5-dibromo-4-hydroxyphenyl)(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzene-1-sulfonate
SMILES [Na+].OC1=C(Br)C=C(C=C1Br)C(=C1C=C(Br)C(=O)C(Br)=C1)C1=CC=CC=C1S([O-])(=O)=O

Specifications

CAS Max % 0.1%, 58.90%, 41%
UN Number UN1170
Grade Chemical Solution
Packaging Poly Bottle
Quantity 100 mL
Chemical Name or Material Bromophenol Blue TS