missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, ACS reagent

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

$47.39 - $272.53

Identifiants chimiques

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
AC403140050
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
403140050
5 g Glass bottle
chaque for $47.39
caisse de 6 chaque for $0.00
Il en reste null
 
AC403140100
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
403140100
10 g Glass bottle
chaque for $83.65
caisse de 6 chaque for $0.00
Il en reste null
 
AC403140500
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
403140500
50 g Glass bottle
chaque for $272.53
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Spécifications

Identifiants chimiques

115-39-9
669.96
UDSAIICHUKSCKT-UHFFFAOYSA-N
8272
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
C19H10Br4O5S
MFCD00005875
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
CHEBI:59424
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Spécifications

279.0°C
Passes Test
MFCD00005875
15, 1454
UDSAIICHUKSCKT-UHFFFAOYSA-N
Authentic
669.96
CHEBI:59424
ACS Reagent
Orange/Pink to Red or Purple
Bromophenol Blue, Pure
115-39-9
C19H10Br4O5S
19, I, 649
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
8272
669.96
from pH 3.0 (yellow) to pH 4.6 (blue)
Powder
Videos
FDS
Documents

Documents

RUO – Research Use Only