missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, ACS reagent

Catalog No. AC403140050
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403140050 5 g Glass bottle
AC403140100 10 g Glass bottle
AC403140500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC403140050 Supplier Thermo Scientific Chemicals Supplier No. 403140050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 279.0°C
Clarity Passes Test
Beilstein 19, I, 649
Merck Index 15, 1454
Infrared Spectrum Authentic
Formula Weight 669.96
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.0 (yellow) to pH 4.6 (blue)
Color Orange/Pink to Red or Purple
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue, Pure
Show More Show Less

RUO – Research Use Only