missing translation for 'onlineSavingsMsg'
Learn More

Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals

Catalog No. AC307050100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307050100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC307050100 Supplier Thermo Scientific Chemicals Supplier No. 307050100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 563-70-2 | Molecular Formula: CH2BrNO2 | Molecular Weight: 139.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 563-70-2
Molecular Formula CH2BrNO2
Molecular Weight (g/mol) 139.94
MDL Number MFCD00007401
InChI Key DNPRVXJGNANVCZ-UHFFFAOYSA-N
Synonym methane, bromonitro, bromo nitro methane, nitrobromomethane, unii-eg63p9eur0, eg63p9eur0, ccris 9456, bromo-nitromethane, 1-bromonitromethane, methane,bromonitro, monobromonitromethane
PubChem CID 68423
IUPAC Name bromo(nitro)methane
SMILES C([N+](=O)[O-])Br

Specifications

Color Yellow
Density 2.0700g/mL
Boiling Point 146°C to 148°C (750.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.495
Linear Formula BrCH2NO2
Quantity 10 g
Specific Gravity 2.07
Formula Weight 139.94
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Bromonitromethane, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the ris
EINECSNumber 209-258-0
RTECSNumber PA5360000
TSCA TSCA

RUO – Research Use Only