missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclopentane 98.0+%, TCI America™

Catalog No. B1006500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1006500G 500 g
B100625G 25 g
B1006100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1006500G Supplier TCI America Supplier No. B1006500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-43-9
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00001359
InChI Key BRTFVKHPEHKBQF-UHFFFAOYSA-N
Synonym cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
PubChem CID 8728
IUPAC Name bromocyclopentane
SMILES C1CCC(C1)Br

Specifications

Color Yellow
Boiling Point 138°C
Quantity 500 g
UN Number 1993
Formula Weight 149.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromocyclopentane
EINECSNumber (3)-4322
TSCA Yes