missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetonitrile 97.0+%, TCI America™

Catalog No. B1058500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1058500G 500 g
B105825G 25 g
B1058100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1058500G Supplier TCI America Supplier No. B1058500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 590-17-0
Molecular Formula C2H2BrN
Molecular Weight (g/mol) 119.95
MDL Number MFCD00001884
InChI Key REXUYBKPWIPONM-UHFFFAOYSA-N
Synonym bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile
PubChem CID 11534
IUPAC Name 2-bromoacetonitrile
SMILES BrCC#N

Specifications

Color Yellow
Boiling Point 150°C
Quantity 500 g
UN Number 3276
Formula Weight 119.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoacetonitrile
RTECSNumber AL7970000
TSCA No