missing translation for 'onlineSavingsMsg'
Learn More

Bromal 96.0+%, TCI America™

Catalog No. T035025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T035025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T035025G Supplier TCI America Supplier No. T035025G

Chemical Identifiers

CAS 115-17-3
Molecular Formula C2HBr3O
Molecular Weight (g/mol) 280.741
MDL Number MFCD00006961
InChI Key YTGSYRVSBPFKMQ-UHFFFAOYSA-N
Synonym tribromoacetaldehyde, bromal, acetaldehyde, tribromo, unii-w2wdi7648e, acetaldehyde,2,2,2-tribromo, acetaldehyde, 2,2,2-tribromo, bromal 6ci, tribromo, tribromoethanal, bromal, redist.
PubChem CID 8256
IUPAC Name 2,2,2-tribromoacetaldehyde
SMILES C(=O)C(Br)(Br)Br

Specifications

Boiling Point 174°C
Quantity 25 g
UN Number 2927
Formula Weight 280.74
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromal
EINECSNumber (2)-2466
RTECSNumber AB3237500
TSCA Yes