missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij(R) C20, Spectrum™ Chemical
BR106, 9004-95-9, C16H33(OCH2CH2)20OH
$254.86 - $4,239.57
Chemical Identifiers
| CAS | 9004-95-9 |
|---|---|
| Molecular Formula | C56H114O21 |
| Molecular Weight (g/mol) | 1123.51 |
| MDL Number | MFCD00080892 |
| InChI Key | NLMKTBGFQGKQEV-UHFFFAOYSA-N |
| IUPAC Name | 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18601064
|
N/A | 25 g | Amber Glass Bottle |
Each for $254.86
|
|
||||
|
18601068
|
N/A | 100 g | Amber Glass Bottle |
Each for $381.18
|
|
||||
|
18601065
|
N/A | 500 g | Amber Glass Bottle |
Each for $717.43
|
|
||||
|
18601066
|
N/A | 2.5 kg | Amber Glass Bottle |
Each for $1,262.75
|
|
||||
|
18601067
|
N/A | 12 kg | Amber Glass Bottle |
Each for $4,239.57
|
|
||||
Description
Spectrum™ Chemical Brij C20 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.
Chemical Identifiers
| 9004-95-9 | |
| 1123.51 | |
| NLMKTBGFQGKQEV-UHFFFAOYSA-N | |
| CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO |
| C56H114O21 | |
| MFCD00080892 | |
| 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol |
Specifications
| 9004-95-9 | |
| C56H114O21 | |
| NLMKTBGFQGKQEV-UHFFFAOYSA-N | |
| 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol | |
| Ungraded |
| 100% | |
| MFCD00080892 | |
| CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO | |
| 1123.51 | |
| Brij 58 |