missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij(R) C20, Spectrum™ Chemical
Description
Spectrum™ Chemical Brij C20 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 100 g |
| Packaging | Amber Glass Bottle |
| Grade | Ungraded |
| Chemical Name or Material | Brij 58 |