missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij(R) C20, Spectrum™ Chemical
BR106, 9004-95-9, C16H33(OCH2CH2)20OH
Supplier: Spectrum Chemical Mfg Cor BR10612KG
| Quantity | 12 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Brij C20 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.
Chemical Identifiers
| 9004-95-9 | |
| 1123.51 | |
| NLMKTBGFQGKQEV-UHFFFAOYSA-N | |
| CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO |
| C56H114O21 | |
| MFCD00080892 | |
| 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol |
Specifications
| Brij 58 | |
| 100% | |
| Amber Glass Bottle | |
| MFCD00080892 | |
| CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO | |
| 1123.51 |
| 9004-95-9 | |
| 12 kg | |
| C56H114O21 | |
| NLMKTBGFQGKQEV-UHFFFAOYSA-N | |
| 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol | |
| Ungraded |